Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Periodic acid 0.6% (w/v) in 80% (v/v) acetic acid

Supplier: Ricca Chemical
Synonyms: Orthoperiodic acidortho-Periodic acid

R5519000-4C
RC55191EA 418.12 USD InStock
RC55191
Periodic acid 0.6% (w/v) in 80% (v/v) acetic acid
Periodic acid


Formula: H₅IO₆
MW: 227.94 g/mol
MDL Number: MFCD00011344
CAS Number: 10450-60-9
UN: 3085
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, strong acetic odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty