Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,10-Phenanthroline 0.1% (w/v) in aqueous solution

Supplier: Ricca Chemical

R5520000-120A R5520000-500A
RC552016EA 91.63 USD InStock
RC552016 RC55204
1,10-Phenanthroline 0.1% (w/v) in aqueous solution
1,10-Phenanthroline


Formula: C₁₂H₈N₂
MW: 180.21 g/mol
MDL Number: MFCD00011678
CAS Number: 66-71-7
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/82/2351982.jpg
Order Now
Specification Test Results
Appearance Clear, light green, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty