Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium potassium sulfate dodecahydrate 10% (w/v)

Supplier: Ricca Chemical
Synonyms: Aluminium Potassium sulphate, 12-HydrateAluminum potassium sulphate dodecahydrateAlum potassiumAluminium Potassium sulfate, 12-HydratePotassium alumAluminium potassium bis(sulfate) dodecahydrateAluminum potassium sulfate dodecahydrateAluminium potassium bis(sulphate) dodecahydrate

R0558000-1A R0558000-500A
RC558-32EA 139.69 USD InStock
RC558-32 RC558-16
Aluminium potassium sulfate dodecahydrate 10% (w/v)
Aluminium potassium sulphate dodecahydrate
Alum, 10% (w/v)


Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Alum Solution APHA (4500-O E)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty