Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol red 1% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

R5725000-500C R5725000-120C
RC5725-16EA 74.47 USD InStock
RC5725-16 RC5725-4
Phenol red 1% (w/v) in aqueous solution indicator
Phenol red


Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Order Now
Specification Test Results
Appearance clear, red, odorless
Suitability for Use Yellow (pH 8.2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty