Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylarsine oxide 0.00564N stabilized analytical reagent

Supplier: Ricca Chemical
Synonyms: OxophenylarsineArzenePAO

R5750000-1C R5750000-4C
RC57501EA 546.04 USD InStock
RC57501 RC5750-32
Phenylarsine oxide 0.00564N stabilized analytical reagent
Phenylarsine oxide
Phenylarsine Oxide (PAO) Titrant (APHA and ASTM)

  • 1mL = 0.2 mg chloride
  • Stabilized


Formula: C₆H₅AsO
MW: 168.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001990
CAS Number: 637-03-6
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.005640 ± 0.000005 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty