Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylarsine oxide 0.0250 N (N/40) stabilized titrant

Supplier: Ricca Chemical
Synonyms: OxophenylarsineArzenePAO

R5755000-1C R5755000-4C
RC575532EA 509.82 USD InStock
RC575532 RC5755-1
Phenylarsine oxide 0.0250 N (N/40) stabilized titrant
Phenylarsine oxide


Formula: C₆H₅AsO
MW: 168.02 g/mol
MDL Number: MFCD00001990
CAS Number: 637-03-6
UN: 3465
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear to hazy, colorless to light tan, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.02500 ± 0.00002 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty