Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylhydrazinium chloride solution 10 g/L

Supplier: Ricca Chemical
Synonyms: Phenylhydrazinium chloridePhenylhydrazine hydrochloride

R5770000-500C
RC5770-16EA 973.68 USD InStock
RC5770-16
Phenylhydrazinium chloride solution 10 g/L
Phenylhydrazinium chloride


Formula: C₆H₉ClN₂
MW: 144.6 g/mol
MDL Number: MFCD00012924
CAS Number: 59-88-1
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/26/2297426.jpg
Order Now
Specification Test Results
Appearance Clear, colorless to slightly yellow, odorless
Assay at 20 °C (traceable to NIST SRM 84) 10.0 ± 0.3 g/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty