Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylhydrazine - sulfuric acid USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R5772000-500C
RC5772-16EA 74.08 USD InStock
RC5772-16
Phenylhydrazine - sulfuric acid USP Test Solution (TS)
Phenylhydrazine - sulfuric acid


Density: 1.5000 g/cm³
Storage Temperature: Ambient
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty