Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phloroglucinol solution 2% (w/v) alcoholic solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: 1,3,5-Trihydroxybenzene

R5775000-500C R5775000-1C
RC5775-32EA 112.64 USD InStock
RC5775-32 RC5775-16
Phloroglucinol solution 2% (w/v) alcoholic solution USP Test Solution (TS)
Phloroglucinol


Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002286
CAS Number: 108-73-6
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, colorless to slight yellow brown, alcohol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty