Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium acetate 10% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Diuretic salt4-Chloro-N-furfuryl-5-sulfamoylanthranlic acidAcetic acid potassium salt

R5866000-120A R5866000-500A
RC5866-16EA 214.41 USD InStock
RC5866-16 RC5866-4
Potassium acetate 10% (w/v) in aqueous solution USP Test Solution (TS)
Potassium acetate


Formula: CH₃COOK
MW: 98.14 g/mol
Boiling Pt: ca.100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
UN: 1789
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/72/16640572.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty