Would you like to log in again?
Potassium Acid Phthalate Standard, 1 ml = 1 mg Chemical Oxygen Demand (COD), 1000 ppm COD, Ricca Chemical Company
Supplier: RICCA Chemical Company
R5868000-500A
RC586816EA 66.53 USD InStock
RC586816
Potassium Acid Phthalate Standard, 1 ml = 1 mg Chemical Oxygen Demand (COD), 1000 ppm COD, Ricca Chemical Company
Standards Environmental Analysis Standards
Standard
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty