Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen phthalate 0.0500 N (N/20)

Supplier: Ricca Chemical
Synonyms: KHPPotassium phthalate monobasicPotassium biphthalatePhthalic Acid Monopotassium SaltPhthalic acid potassium salt

R5870000-1A R5870000-500A
RC587016EA 82.8 USD InStock
RC587016 RC587032
Potassium hydrogen phthalate 0.0500 N (N/20)
Potassium hydrogen phthalate


Formula: HOOCC₆H₄COOK
MW: 204.22 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0043 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/58/16640658.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.05000 ± 0.00001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty