Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony(III) potassium oxitartrate 0.2743% (1.3715 g/500 mL) (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning
Synonyms: Antimony potassium L(+)-tartrate(+)-Antimony potassium L-tartrate(R,R)-(+)-Antimony potassium tartrateL(+)-Tartaric acid antimony potassium saltPotassium Antimonyl Tartrate(R,R)-(+)-Tartaric acid antimony potassium saltL(+)-Antimony potassium tartrate

R5872000-1A R5872000-500A
RC587216EA 86.49 USD InStock
RC587216 RC587232
Antimony(III) potassium oxitartrate 0.2743% (1.3715 g/500 mL) (w/v) in aqueous solution
Antimony (III) potassium oxitartrate


Formula: C₈H₄K₂O₁₂Sb₂
MW: 613.83 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Refrigerator
CAS Number: 11071-15-1
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Certified Concentration 1.3715 g/ 500 mL

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty