Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride saturated ∼4 M

Supplier: Ricca Chemical
Synonyms: Potassiumchloride

R5900000-4A R5900000-120A R5900000-500A R5900000-1A
RC5900-1EA 218.58 USD InStock
RC5900-1 RC59004 RC590016 RC590032
Potassium chloride saturated ∼4 M
Potassium chloride


Formula: KCl
MW: 74.55 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1700 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty