Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hexachloroplatinate(IV) 0.2% (w/v) in hydrochloric acid 10%

Supplier: Ricca Chemical
Synonyms: Dipotassium hexachloroplatinate

R5950000-120C R5950000-500C
RC5950-16EA 415.47 USD InStock
RC5950-16 RC5950-4
Potassium hexachloroplatinate(IV) 0.2% (w/v) in hydrochloric acid 10%
Potassium hexachloroplatinate(IV)


Formula: K₂PtCl₆
MW: 485.99 g/mol
MDL Number: MFCD00011389
CAS Number: 16921-30-5
UN: 3290
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear, light yellow, odorless to slightly pungent odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty