Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chromate 0.063% (w/v) in aqueous solution

Supplier: Ricca Chemical

R5990000-1A R5990000-500A R5990000-120A
RC599016EA 32.74 USD InStock
RC599016 RC59904 RC5990-32
Potassium chromate 0.063% (w/v) in aqueous solution
Potassium chromate


Formula: K₂CrO₄
MW: 194.19 g/mol
MDL Number: MFCD00011368
CAS Number: 7789-00-6
UN: 3087
ADR: 5.1,III
Order Now
Specification Test Results
Appearance Clear, yellow, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty