Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chromate 5% (w/v) in aqueous solution indicator, for argentometric titration chloride-free

Supplier: Ricca Chemical

R6000000-500A R6000000-4A R6000000-1A R6000000-120A
RC600016EA 66.9 USD InStock
RC600016 RC600032 RC60004 RC6000-1
Potassium chromate 5% (w/v) in aqueous solution indicator, for argentometric titration chloride-free
Potassium chromate


Formula: K₂CrO₄
MW: 194.19 g/mol
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3287
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, yellow, odorless
Suitability for Use Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty