Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chromate 10% (w/v) in aqueous solution USP Test Solution (TS) indicator, for argentometric titration chloride-free

Supplier: Ricca Chemical
Danger

R6005000-1A R6005000-500A
RC6005-16EA 110.27 USD InStock
RC6005-16 RC6005-32
Potassium chromate 10% (w/v) in aqueous solution USP Test Solution (TS) indicator, for argentometric titration chloride-free
Potassium chromate


Formula: K₂CrO₄
MW: 194.19 g/mol
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3287
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, yellow, odorless
Refractive Index 10.0 ± 0.3 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty