Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium cyanide 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

R6040000-500B
RC604016EA 132.02 USD InStock
RC604016
Potassium cyanide 10% (w/v) in aqueous solution
Potassium cyanide


Formula: KCN
MW: 65.12 g/mol
MDL Number: MFCD00011397
CAS Number: 151-50-8
UN: 1680
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear, colorless, cyanide odor (do not smell directly)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty