Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia, Dilute R1 102 g/L

Supplier: Ricca Chemical
Danger

R0610200-500A
RC610.2-16EA 125.53 USD InStock
RC610.2-16
Ammonia, Dilute R1 102 g/L
Ammonia, Dilute R1


Formula: H3N
MW: 17,03 g/mol
Boiling Pt: ~ 36 °C
Melting Pt: ~ 0 °C
Density: 0.9555 g/cm³
Storage Temperature: Ambient
UN: 2672
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, ammonia odor
Assay at 20 °C (traceable to NIST SRM 723) 102 ± 2 g/L NH3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty