Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium fluoride 40% (w/v) in aqueous solution

Supplier: Ricca Chemical

R6100000-1A R6100000-500A
RC610016EA 171.51 USD InStock
RC610016 RC610032
Potassium fluoride 40% (w/v) in aqueous solution
Potassium fluoride


Formula: KF
MW: 58.1 g/mol
MDL Number: MFCD00011398
CAS Number: 7789-23-3
UN: 1812
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 8.30 ± 0.10 pH at 25.0 °C
Specific Gravity 1.20 ± 0.01 at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty