Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amine sulfuric acid reagent for sulfide determination (methylene blue method)

Supplier: Ricca Chemical
Danger

R0610000-1C1 R0610000-500C1
RC61032EA 283.9 USD InStock
RC61032 RC61016
Amine sulfuric acid reagent for sulfide determination (methylene blue method)
Amine sulfuric acid reagent


Formula: H2O4S
MW: 98.072 g/mol
Density: 1.5000 g/cm³
Storage Temperature: Ambient
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Pale yellow to slight tan liquid, odorless
Suitability for Use Pink to Clear, colorless when Sulfide free

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty