Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 20% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger
Synonyms: Potash causticPotassium hydrateCaustic potash

R6140000-250A R6140000-500A R6140000-4A R6140000-1A
RCR6140000250AEA 81.33 USD InStock
RCR6140000250A RC6140-1 RC614016 RC6140-32
Potassium hydroxide 20% (w/v) in aqueous solution
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Boiling Pt: ~100 °C
Melting Pt: −13 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 19.94 - 20.06 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty