Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammoniacal potassium ferricyanide USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R0617500-500A
RC617.5-16EA 150.53 USD InStock
RC617.5-16
Ammoniacal potassium ferricyanide USP Test Solution (TS)
Ammoniacal potassium ferricyanide


Boiling Pt: ~ 36 °C
Density: 0.9950 g/cm³
Storage Temperature: Ambient
UN: 3266
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, yellow, Ammonia odor
Suitability for Use Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty