Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium acetate 10% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Acetic acid ammonium salt

R0618000-1A R0618000-500A
RC618-16EA 60.2 USD InStock
RC618-16 RC618-32
Ammonium acetate 10% (w/v) in aqueous solution USP Test Solution (TS)
Ammonium acetate


Formula: H₃CCOONH₄
MW: 77.08 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0200 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/04/16640604.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Specific Gravity 10.0 ± 1.2 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty