Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 0.0100 N (N/100)

Supplier: Ricca Chemical
Synonyms: Potash causticPotassium hydrateCaustic potash

R6196000-500A R6196000-1A
RC6196-32EA 70.86 USD InStock
RC6196-32 RC6196-16
Potassium hydroxide 0.0100 N (N/100)
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.01000 ± 0.00005 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty