Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium carbonate solution R 158 g/L

Supplier: Ricca Chemical
Warning

R0622150-500A
RC622.15-16EA 162.01 USD InStock
RC622.15-16
Ammonium carbonate solution R 158 g/L
Ammonium carbonate solution R


Order Now
Specification Test Results
Appearance Clear, colorless, ammonia odor
Assay at 20 °C (traceable to NIST SRM 723) 158.0 ± 0.8 g/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty