Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium chloride 1.15 g/L for BOD determination pH 7.20

Supplier: Ricca Chemical
Warning
Synonyms: Ammonium muriateSalmiac

R0623000-500A R0623000-120A
RC6234EA 31.34 USD InStock
RC6234 RC62316
Ammonium chloride 1.15 g/L for BOD determination pH 7.20
Ammonium chloride


Formula: NH₄Cl
MW: 53.49 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0004 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/71/16640571.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.20 ± 0.05 pH at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty