Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium chloride solution R 10.7% (w/v) in water

Supplier: Ricca Chemical
Danger
Synonyms: Ammonium muriateSalmiac

R0626300-500A
RC626.3-16EA 130.32 USD InStock
RC626.3-16
Ammonium chloride solution R 10.7% (w/v) in water
Ammonium chloride


Formula: NH₄Cl
MW: 53.49 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0320 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 107.0 ± 0.5 g/L NH4Cl

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty