Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 1.00 N in denatured alcohol

Supplier: Ricca Chemical
Danger
Synonyms: Potassium hydrateCaustic potash

R6266900-1A R6266900-4A
RC6266.9-1EA 514.53 USD InStock
RC6266.9-1 RC6266.9-32
Potassium hydroxide 1.00 N in denatured alcohol
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, colorless, alcohol odor
Assay at 20 °C (traceable to NIST SRM 723) 1.000 ± 0.005 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty