Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium iodate 0.0500 N (N/20)

Supplier: Ricca Chemical

R6274000-1C
RC6274-32EA 92.46 USD InStock
RC6274-32
Potassium iodate 0.0500 N (N/20)
Potassium iodate


Formula: KlO₃
MW: 214 g/mol
MDL Number: MFCD00011406
CAS Number: 7758-05-6
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.05000 ± 0.00001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty