Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium iodate-iodide solution 0.0125 N (N/80)

Supplier: Ricca Chemical
Warning

R6280000-20F R6280000-4C R6280000-1C
RC62805EA 462.02 USD InStock
RC62805 RC628032 RC62801
Potassium iodate-iodide solution 0.0125 N (N/80)
Potassium iodate/-iodide, 0.713g/l KIO3 (0.02 N) aqueous solution
1 ml = 0.5 mg SO3. APHA, for Sulfite.


Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.01250 ± 0.00001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty