Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dipotassium oxalate 2% (w/v) in aqueous solution

Supplier: Ricca Chemical

R6320000-500A R6320000-1A
RC6320-32EA 85.57 USD InStock
RC6320-32 RC632016
Dipotassium oxalate 2% (w/v) in aqueous solution
di-Potassium oxalate


Formula: C₂K₂O₄
MW: 166.22 g/mol
CAS Number: 583-52-8
UN: 3282
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index 2.0 ± 0.3 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty