Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyridine-Barbituric acid APHA for cyanide determination

Supplier: Ricca Chemical
Danger

6584-16 6584-32
RC6584-32EA 263.58 USD InStock
RC6584-32 RC6584-16
Pyridine-Barbituric acid APHA for cyanide determination
Pyridine-Barbituric acid
This reagent contains 6% (w/v) Reagent Grade Barbituric Acid, CAS No. 67-52-7, 30% (v/v) ACS Reagent Grade Pyridine, CAS No. 110-86-1, and 6% (v/v) ACS Reagent Grade Hydrochloric Acid, CAS No. 7647-01-0, in ACS Reagent Grade Water, CAS No. 7732-18-5.


Storage Temperature: Refrigerator
UN: 2924
ADR: 3 (8),III
Order Now
Specification Test Results
Appearance Golden yellow liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty