Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium heptamolybdate 50 g/L in hydrochloric acid 16.7%

Supplier: Ricca Chemical
Danger

R0662000-1A
RC66232EA 338.71 USD InStock
RC66232
Ammonium heptamolybdate 50 g/L in hydrochloric acid 16.7%
Ammonium heptamolybdate solutions


Formula: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235.86 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0700 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty