Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium heptamolybdate 4% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning

R0665000-1A R0665000-4A R0665000-500A
RC665-32EA 160.18 USD InStock
RC665-32 RC66516 RC665-1
Ammonium heptamolybdate 4% (w/v) in aqueous solution
Ammonium heptamolybdate solutions


Formula: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235.86 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0400 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
Order Now
Specification Test Results
Appearance Clear, colorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty