Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium heptamolybdate 7.5% (w/v) in aqueous solution, pH adjusted

Supplier: Ricca Chemical
Danger

R0667000-1A R0667000-500A
RC667-32EA 190.68 USD InStock
RC667-32 RC66716
Ammonium heptamolybdate 7.5% (w/v) in aqueous solution, pH adjusted
Ammonium heptamolybdate solutions


Formula: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235.86 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0700 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.5 ± 0.5 pH at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty