Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safranine O alcoholic solution counterstain for gram staining

Supplier: Ricca Chemical
Danger
Synonyms: Safranin TSafranin Y3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chlorideSafranin OGossypimine3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chlorideCI 50240Basic Red 2Cotton RedSafranin ASafranin

R6680000-500A R6680000-4A R6680000-1A
RC668032EA 125.23 USD InStock
RC668032 RC668016 RC6680-1
Safranine O alcoholic solution counterstain for gram staining
Safranine O


Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 0.9000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Clear, dark red, slight alcohol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty