Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver diethyldithiocarbamate 0.3% (w/v) in chloroform

Supplier: Ricca Chemical
Synonyms: Diethyldithiocarbamic acid silver saltN,N-Diethyldithiocarbamic acid silver salt(Diethyldithiocarbamato-S,S')silverDETC

R6810000-500C R6810000-120C
RC6810-16EA 278.08 USD InStock
RC6810-16 RC6810-4
Silver diethyldithiocarbamate 0.3% (w/v) in chloroform
Silver diethyldithiocarbamate


Formula: (C₂H₅)₂NCSSAg
MW: 256.14 g/mol
MDL Number: MFCD00004929
CAS Number: 1470-61-7
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Greenish-yellow, chloroform odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty