Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diammonium hydrogen orthophosphate 30% (w/v) in aqueous solution (1 ml = 1 mg Phenol) APHA for magnesium

Supplier: Ricca Chemical
Warning
Synonyms: Ammonium Phosphatedi-Ammonium hydrogen phosphateDiammonium hydrogen orthophosphateAmmonium hydrogen phosphateAmmonium monohydrogen phosphateAmmonium phosphate dibasic

R0689300-1A R0689300-500A
RC689.30-32EA 179.7 USD InStock
RC689.30-32 RC689.30-16
Diammonium hydrogen orthophosphate 30% (w/v) in aqueous solution (1 ml = 1 mg Phenol) APHA for magnesium
di-Ammonium hydrogen orthophosphate


Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Density: 1.2000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
Order Now
Specification Test Results
Appearance Clear, colorless liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty