Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diammonium hydrogen orthophosphate solution 400 g/800 mL in water

Supplier: Ricca Chemical
Warning
Synonyms: di-Ammonium hydrogen phosphate Diammonium hydrogen orthophosphate Ammonium monohydrogen phosphate Ammonium phosphate dibasic Ammonium hydrogen phosphate

R0690000-500A
RC69016EA 342.46 USD InStock
RC69016
Diammonium hydrogen orthophosphate solution 400 g/800 mL in water
di-Ammonium hydrogen orthophosphate


Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Density: 1.2000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
Order Now
Specification Test Results
Appearance Clear, colorless, odorless to slight ammonia odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty