Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate 0.0192 M (0.0192 N)

Supplier: Ricca Chemical
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

R6910000-1B R6910000-4B
RC691032EA 101.26 USD InStock
RC691032 RC6910-1
Silver nitrate 0.0192 M (0.0192 N)
Silver nitrate
1 mL = 1 mg CN-


Formula: AgNO₃
MW: 169.87 g/mol
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.01920 ± 0.00001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty