Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium thiocyanate 30% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger
Synonyms: Ammonium rhodanide

R0698000-500A R0698000-4A R0698000-1A
RC698-16EA 159.84 USD InStock
RC698-16 RC698-32 RC698-1
Ammonium thiocyanate 30% (w/v) in aqueous solution
Ammonium thiocyanate


Formula: NH₄SCN
MW: 76.12 g/mol
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Suitability for Use Red color with Iron (Fe)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty