Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium metavanadate USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Ammonium vanadium oxideAmmonium monovanadateAmmonium vanadium trioxide

R0698300-500A
RC698.3-16EA 80.67 USD InStock
RC698.3-16
Ammonium metavanadate USP Test Solution (TS)
Ammonium metavanadate


Formula: NH₄VO₃
MW: 116.98 g/mol
MDL Number: MFCD00011430
CAS Number: 7803-55-6
UN: 2859
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear, yellow, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty