Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate 0.100 N (N/10) in denatured alcohol

Supplier: Ricca Chemical
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

R7028000-4B R7028000-1B
RC7028-32EA 183.89 USD InStock
RC7028-32 RC7028-1
Silver nitrate 0.100 N (N/10) in denatured alcohol
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, alcohol odor
Assay at 20 °C (traceable to NIST SRM 999) 0.1000 ± 0.0001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty