Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate R1 Reag. Ph. Eur. , 42.5 g/L (0.250 N)

Supplier: Ricca Chemical
Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

R7040000-500C
RC7040-16EA 241.47 USD InStock
RC7040-16
Silver nitrate R1 Reag. Ph. Eur. , 42.5 g/L (0.250 N)
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0400 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 42.50 ± 0.02 g/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty