Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Simulated gastric fluid (without pepsin) (0.2% NaCl) (w/v) in hydrochloric acid 0.7%

Supplier: Ricca Chemical
Danger

R7108000-1A R7108000-10F R7108000-20F R7108000-500A
RC7108-16EA 91.09 USD InStock
RC7108-16 RC7108-32 RC7108-2.5 RC7108-5
Simulated gastric fluid (without pepsin) (0.2% NaCl) (w/v) in hydrochloric acid 0.7%
Simulated gastric fluid (without pepsin)


Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.1188 ± 0.0050 N at 20.0 °C
pH at 25 °C (traceable to NIST SRM 185 & 186) 1.2 ± 0.2 pH at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty