Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Simulated intestinal fluid USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R7109000-1A R7109000-500A
RC7109-16EA 133.05 USD InStock
RC7109-16 RC7109-32
Simulated intestinal fluid USP Test Solution (TS)
Simulated intestinal fluid


Order Now
Specification Test Results
Appearance Clear to hazy with stringy fibers, colorless, characteristic odor
pH at 25 °C (traceable to NIST SRM 185 & 186) 6.8 ± 0.1 pH at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty