Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium metaarsenite 0.5% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7140000-500A R7140000-1A
RC714032EA 76.94 USD InStock
RC714032 RC714016
Sodium metaarsenite 0.5% (w/v) in aqueous solution
Sodium metaarsenite


Formula: NaAsO₂
MW: 129.91 g/mol
MDL Number: MFCD00003472
CAS Number: 7784-46-5
UN: 2027
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty