Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium metaarsenite 0.100 M (M/10)

Supplier: Ricca Chemical

R7142000-1C R7142000-500C
RC7142-32EA 108.77 USD InStock
RC7142-32 RC7142-16
Sodium metaarsenite 0.100 M (M/10)
Sodium metaarsenite


Formula: NaAsO₂
MW: 129.91 g/mol
MDL Number: MFCD00003472
CAS Number: 7784-46-5
UN: 2027
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 83) 0.1000 ± 0.0001 M at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty